Assay Details
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016074.4 | NP_057158.1 | 1 - 2 | 93 | 92 | |
GenBank mRNA | BC002771.2 | - | 1 - 2 | 53 | 92 | |
BC063405.1 | - | 1 - 2 | 53 | 5190565 | 92 | |
CA389141.1 | - | 1 - 2 | 73 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01006327.hg
Additional Information:
This assay is in the 5'UTR region of the BC063405.1,NM_016074.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01018017.hg.1 | TC01006327.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018017.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018017.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018017.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018014.hg.1 | TC01006327.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018014.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018014.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018014.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018014.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018019.hg.1 | TC01006327.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018019.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018019.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018019.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018019.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018018.hg.1 | TC01006327.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018018.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018018.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018018.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171276.hg.1 | TC0100009881.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171275.hg.1 | TC0100009881.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171272.hg.1 | TC0100009881.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009881.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369150.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369152.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000369153.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | ENST00000476344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | NM_016074.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171277.hg.1 | TC0100009881.hg.1 | Clariom D TC |
