Assay Details
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286694.1 | NP_001273623.1 | 2 - 3 | 1007 | 139 | |
NM_033429.2 | NP_219501.2 | 3 - 4 | 1038 | 139 | ||
GenBank mRNA | AK296804.1 | - | 3 - 4 | 260 | 139 | |
BC009516.2 | - | 2 - 3 | 214 | 139 | ||
DB204573.1 | - | 3 - 4 | 262 | 139 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001591.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001286694.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000354678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000395465.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000448060.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000467889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000478113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | ENST00000540479.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001031733.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_001286695.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NM_033429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010886.hg.1 | NR_104583.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 185294 | - | - |
