Assay Details
| Gene Symbol: STAMBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020799.3 | NP_065850.1 | 1 - 2 | 445 | 102 | |
| XM_006717928.2 | XP_006717991.1 | 2 - 3 | 546 | 102 | ||
| XM_017016457.1 | XP_016871946.1 | 1 - 2 | 792 | 102 | ||
| GenBank mRNA | AB037794.1 | - | 1 - 2 | 766 | 102 | |
| AK056086.1 | - | 1 - 2 | 374 | 102 | ||
| BC010846.1 | - | 1 - 2 | 363 | 3898533 | 102 | |
| CD244526.1 | - | 1 - 2 | 470 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002941.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK056086.1,BC010846.1,NM_020799.3,XM_006717928.2,XM_017016457.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10006941.hg.1 | TC10002941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006941.hg.1 | ENST00000371924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006941.hg.1 | ENST00000371926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006941.hg.1 | ENST00000371927.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006941.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006940.hg.1 | TC10002941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006940.hg.1 | ENST00000371924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006940.hg.1 | ENST00000371926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006940.hg.1 | ENST00000371927.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006940.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006932.hg.1 | TC10002941.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006932.hg.1 | ENST00000371926.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006932.hg.1 | ENST00000371927.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006932.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10006932.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156806.hg.1 | TC1000008385.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156795.hg.1 | TC1000008385.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371924.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | ENST00000468698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008385.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156796.hg.1 | TC1000008385.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000371926.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000371927.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | ENST00000461650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | NM_020799.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000156797.hg.1 | TC1000008385.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124733 | - | - |

STAMBPL1
Genomic Map