Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005519.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005519.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005519.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005519.hg.1 |
TC10000470.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008047.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
ENST00000372833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
ENST00000372837.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
NM_203298.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000154573.hg.1 |
TC1000008047.hg.1 |
Clariom D TC |