Assay Details
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164522.2 | NP_001157994.1 | 2 - 3 | 280 | 62 | |
NM_001304839.1 | NP_001291768.1 | 2 - 3 | 280 | 62 | ||
NM_177403.5 | NP_796377.3 | 1 - 2 | 354 | 62 | ||
XM_006711288.3 | XP_006711351.1 | 2 - 3 | 277 | 62 | ||
GenBank mRNA | AK122657.1 | - | 2 - 3 | 275 | 62 | |
AY094596.1 | - | 2 - 3 | 227 | 62 | ||
BC017092.1 | - | 2 - 3 | 222 | 3851998 | 62 | |
BI757251.1 | - | 1 - 2 | 351 | 62 | ||
DA966224.1 | - | 2 - 3 | 280 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
protein transport
positive regulation of interleukin-6 production
negative regulation of toll-like receptor 4 signaling pathway
negative regulation of toll-like receptor 9 signaling pathway
late endosome to Golgi transport
positive regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
GTP binding
small GTPase mediated signal transduction
protein transport
positive regulation of interleukin-6 production
negative regulation of toll-like receptor 4 signaling pathway
negative regulation of toll-like receptor 9 signaling pathway
late endosome to Golgi transport
positive regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
GTP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229518.hg.1 | TC0100017083.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100229521.hg.1 | TC0100017083.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617070.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000617991.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000622969.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623597.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | ENST00000623893.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001164522.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_001304839.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017083.hg.1 | NM_177403.5 | Refseq |
