Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250.5 | NP_001241.1 | 1 - 2 | 143 | 102 | |
| NM_001302753.1 | NP_001289682.1 | 1 - 2 | 143 | 102 | ||
| NM_001322421.1 | NP_001309350.1 | 1 - 2 | 143 | 102 | ||
| NM_001322422.1 | NP_001309351.1 | 1 - 2 | 143 | 102 | ||
| NM_152854.3 | NP_690593.1 | 1 - 2 | 143 | 102 | ||
| NR_126502.1 | 1 - 2 | 143 | 102 | |||
| NR_136327.1 | 1 - 2 | 143 | 102 | |||
| XM_005260619.3 | XP_005260676.1 | 1 - 2 | 155 | 102 | ||
| XM_011529109.2 | XP_011527411.1 | 1 - 2 | 155 | 102 | ||
| XM_017028135.1 | XP_016883624.1 | 1 - 2 | 155 | 102 | ||
| XM_017028136.1 | XP_016883625.1 | 1 - 2 | 154 | 102 | ||
| XM_017028137.1 | XP_016883626.1 | 1 - 2 | 152 | 102 | ||
| GenBank mRNA | AJ300189.1 | - | 1 - 2 | 81 | 102 | |
| AK222896.1 | - | 1 - 2 | 83 | 102 | ||
| BC012419.1 | - | 1 - 2 | 110 | 3856903 | 102 | |
| BC064518.1 | - | 1 - 2 | 119 | 5227115 | 102 | |
| BM761221.1 | - | 1 - 2 | 130 | 102 | ||
| BX364285.2 | - | 1 - 2 | 97 | 102 | ||
| HY063089.1 | - | 1 - 2 | 143 | 102 | ||
| HY088000.1 | - | 1 - 2 | 143 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ300189.1, BC012419.1, BC064518.1, NM_001250.5, NM_001302753.1, NM_001322421.1, NM_001322422.1, NM_152854.3, XM_005260619.3, XM_011529109.2, XM_017028135.1, XM_017028136.1, XM_017028137.1 transcripts.
Set Membership: |
|

CD40
Genomic Map