Assay Details
| Gene Symbol: RACGAP1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026583.1 | 1 | 47 | 93 | ||
| GenBank mRNA | AF334184.1 | - | 1 | 47 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001418.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF334184.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16553873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758365 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16553899 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323900 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323883 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758357 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323892 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758388 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758411 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758408 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323890 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16553895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758410 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758399 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323901 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758403 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758412 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28758379 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08323904 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 38623 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 1267 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 364929 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P65041 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P65041 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 33981 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P201437 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P134441 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P134441 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 11842 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P65041 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P65041 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16553895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16553899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758407 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323894 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758404 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323902 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323898 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16553895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323907 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323892 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323895 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758405 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323901 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323904 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323890 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323881 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758402 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758399 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758388 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758408 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323918 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323889 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16553899 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08323887 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28758379 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P65041 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P201437 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P134441 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P201437 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P134441 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P201437 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P134441 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P65041 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 19935 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 6938 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 85555 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P134441 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323912 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323883 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323910 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323877 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323908 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323903 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323919 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323897 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323905 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323925 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16553893 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323881 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323899 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323921 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323896 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323867 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323890 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323879 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323904 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323923 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323927 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323914 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323900 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323916 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323888 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323894 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323878 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08323891 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P134441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7962488 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7962487 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224272_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR12018368.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12018368.hg.1 | TC12001418.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010494.hg.1 | ENST00000550663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179141.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179141.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179141.hg.1 | TC1200010494.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010494.hg.1 | ENST00000550663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010494.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179140.hg.1 | ENST00000550663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200179140.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179140.hg.1 | ENST00000550663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200179140.hg.1 | NR_026583.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200179140.hg.1 | TC1200010494.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 224272_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224272_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224272_at7 | - | - |
| Human Genome U133 Set | 224272_at | - | - |
| Human Genome U133 Set | 224272_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047498 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 154565 | - | - |

RACGAP1P
Genomic Map