Assay Details
Gene Symbol: LOC171391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF435951.1 | - | 1 | 156 | 72 | |
AK314117.1 | - | 1 | 657 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P412427 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 3373 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P111631 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P111631 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30761 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P412427 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P412427 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P111631 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P111631 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 10189 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7937475 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7937474 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1553943_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000533938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000533938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000623692.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000623846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | ENST00000623846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000525941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000526588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000530083.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000533938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000533938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000623692.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000623846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | ENST00000623846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126342.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006488.hg.1 | NR_126343.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1553943_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553943_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1553943_at8 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148033 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3305980 | - | - |
