Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320972.1 | NP_001307901.1 | 1 - 2 | 460 | 92 | |
| NM_001320973.1 | NP_001307902.1 | 1 - 2 | 460 | 92 | ||
| NM_003666.3 | NP_003657.1 | 1 - 2 | 460 | 92 | ||
| XM_005245561.4 | XP_005245618.1 | 1 - 2 | 367 | 92 | ||
| XM_017002645.1 | XP_016858134.1 | 1 - 2 | 330 | 92 | ||
| GenBank mRNA | AF288161.1 | - | 1 - 2 | 350 | 92 | |
| AK223533.1 | - | 1 - 2 | 322 | 92 | ||
| AK291725.1 | - | 1 - 2 | 127 | 92 | ||
| AK307151.1 | - | 1 - 2 | 132 | 92 | ||
| BC020716.1 | - | 1 - 2 | 109 | 4723421 | 92 | |
| BI458959.1 | - | 1 - 2 | 384 | 92 | ||
| BI460514.1 | - | 1 - 2 | 375 | 92 | ||
| U79751.1 | - | 1 - 2 | 349 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001475.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF288161.1,AK291725.1,BC020716.1,NM_001320972.1,NM_001320973.1,NM_003666.3,U79751.1,XM_005245561.4,XM_017002645.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of transcription from RNA polymerase II promoter
Golgi organization
cell proliferation
vesicle-mediated transport
Golgi to plasma membrane protein transport
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
ubiquitin protein ligase binding
regulation of cell growth
regulation of transcription from RNA polymerase II promoter
Golgi organization
cell proliferation
vesicle-mediated transport
Golgi to plasma membrane protein transport
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01022858.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022858.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022858.hg.1 | ENST00000367807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022858.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022858.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022856.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022856.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022856.hg.1 | ENST00000367807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022856.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022856.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022861.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022861.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022861.hg.1 | ENST00000367807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022861.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022861.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022862.hg.1 | TC01001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022862.hg.1 | ENST00000329281.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022862.hg.1 | ENST00000367807.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022862.hg.1 | ENST00000367808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022862.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177612.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177615.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177616.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177617.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100177610.hg.1 | TC0100010546.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000329281.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367807.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000367808.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000420531.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | ENST00000426663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010546.hg.1 | NM_003666.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 136933 | - | - |

BLZF1
Genomic Map