| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000329281.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367807.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000367808.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000420531.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
ENST00000426663.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010546.hg.1 |
NM_003666.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009528.hg.1 |
ENST00000553776.1 |
Ensembl |