Assay Details
| Gene Symbol: SIRPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040022.1 | NP_001035111.1 | 7 - 8 | 1589 | 69 | |
| NM_001040023.1 | NP_001035112.1 | 6 - 7 | 1497 | 69 | ||
| NM_080792.2 | NP_542970.1 | 7 - 8 | 1256 | 69 | ||
| GenBank mRNA | AB023430.1 | - | 7 - 8 | 1256 | 69 | |
| AK290776.1 | - | 7 - 8 | 1362 | 69 | ||
| AK298243.1 | - | 7 - 8 | 1207 | 69 | ||
| AK312521.1 | - | 6 - 7 | 1310 | 69 | ||
| BC038510.2 | - | 6 - 7 | 1494 | 5261789 | 69 | |
| BC075849.1 | - | 7 - 8 | 1435 | 4589732 | 69 | |
| D86043.1 | - | 6 - 7 | 1228 | 69 | ||
| Y10375.1 | - | 6 - 7 | 1238 | 69 | ||
| Y11047.1 | - | 5 - 6 | 1004 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000019.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | ENST00000356025.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | ENST00000400068.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20000232.hg.1 | TC20000019.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | TC2000006501.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144539.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000006501.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_080792.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | TC2000006501.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000356025.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000358771.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000400068.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | ENST00000622179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_001040022.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_001040023.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000144540.hg.1 | NM_080792.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 183616 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 219592 | - | - |

SIRPA
Genomic Map