Assay Details
Gene Symbol: RABEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291581.1 | NP_001278510.1 | 15 - 16 | 2461 | 155 | |
NM_004703.5 | NP_004694.2 | 16 - 17 | 2590 | 155 | ||
XM_011524060.1 | XP_011522362.1 | 17 - 18 | 2656 | 155 | ||
XM_017025294.1 | XP_016880783.1 | 15 - 16 | 2183 | 155 | ||
XM_017025295.1 | XP_016880784.1 | 14 - 15 | 2117 | 155 | ||
GenBank mRNA | AK297535.1 | - | 15 - 16 | 2450 | 155 | |
AK314183.1 | - | 16 - 17 | 2581 | 155 | ||
BC041700.1 | - | 16 - 17 | 2556 | 6046320 | 155 | |
BX103338.1 | - | 3 - 4 | 341 | 155 | ||
X77723.1 | - | 16 - 17 | 2439 | 155 | ||
X91141.1 | - | 16 - 17 | 2554 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
