Assay Details
| Gene Symbol: POLR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007055.3 | NP_008986.2 | 26 - 27 | 3567 | 73 | |
| GenBank mRNA | AF021351.1 | - | 27 - 28 | 3434 | 73 | |
| AK026023.1 | - | 17 - 18 | 2141 | 73 | ||
| AK300329.1 | - | 27 - 28 | 3484 | 73 | ||
| AY091461.1 | - | 11 - 12 | 1241 | 73 | ||
| AY091462.1 | - | 2 - 3 | 161 | 73 | ||
| BC014399.1 | - | 3 - 4 | 273 | 73 | ||
| BC041089.1 | - | 26 - 27 | 3528 | 3915501 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001446.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC014399.1 transcripts.
Set Membership: |
|

POLR3A
Genomic Map