Assay Details
| Gene Symbol: SEC16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276418.1 | NP_001263347.1 | 25 - 26 | 6878 | 124 | |
| NM_014866.1 | NP_055681.1 | 26 - 27 | 6868 | 124 | ||
| XM_005266134.2 | XP_005266191.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519246.2 | XP_011517548.1 | 26 - 27 | 6877 | 124 | ||
| XM_011519247.1 | XP_011517549.1 | 25 - 26 | 6734 | 124 | ||
| XM_011519248.2 | XP_011517550.1 | 25 - 26 | 6708 | 124 | ||
| XM_011519249.1 | XP_011517551.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519250.2 | XP_011517552.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519251.1 | XP_011517553.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519252.2 | XP_011517554.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519253.1 | XP_011517555.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519254.1 | XP_011517556.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519255.2 | XP_011517557.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519256.1 | XP_011517558.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519257.1 | XP_011517559.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519258.1 | XP_011517560.1 | 24 - 25 | 6590 | 124 | ||
| XM_011519259.1 | XP_011517561.1 | 25 - 26 | 6731 | 124 | ||
| XM_011519260.2 | XP_011517562.1 | 25 - 26 | 6715 | 124 | ||
| XM_011519261.1 | XP_011517563.1 | 22 - 23 | 6284 | 124 | ||
| XM_011519262.1 | XP_011517564.1 | 26 - 27 | 6020 | 124 | ||
| XM_011519263.2 | XP_011517565.1 | 26 - 27 | 6020 | 124 | ||
| XM_017015363.1 | XP_016870852.1 | 25 - 26 | 6731 | 124 | ||
| GenBank mRNA | AB002308.4 | - | 26 - 27 | 6868 | 124 | |
| AB593115.1 | - | 25 - 26 | 6878 | 124 | ||
| AK074565.1 | - | 1 - 2 | 93 | 124 | ||
| AK094756.1 | - | 4 - 5 | 840 | 124 | ||
| AK301371.1 | - | 3 - 4 | 198 | 124 | ||
| AK302210.1 | - | 2 - 3 | 134 | 124 | ||
| AK316194.1 | - | 1 - 2 | 93 | 124 | ||
| BC001404.2 | - | 9 - 10 | 947 | 124 | ||
| BC008332.1 | - | 15 - 16 | 1741 | 124 | ||
| BC028183.1 | - | 21 - 22 | 2735 | 124 | ||
| BC098454.1 | - | 9 - 10 | 1031 | 124 | ||
| BC125018.1 | - | 24 - 25 | 5440 | 124 | ||
| BC125019.1 | - | 25 - 26 | 4729 | 124 | ||
| DQ903855.1 | - | 24 - 25 | 6025 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001732.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074565.1,AK301371.1,AK302210.1,AK316194.1 transcripts.
Set Membership: |
|

SEC16A
Genomic Map