Assay Details
| Gene Symbol: KHNYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290256.1 | NP_001277185.1 | 1 - 2 | 123 | 115 | |
| GenBank mRNA | AB002321.1 | - | 1 - 2 | 123 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146881.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146881.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146881.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146881.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146881.hg.1 | TC1400006742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000554268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006742.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000251343.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000553935.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000556510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | ENST00000556842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_001290256.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_001290257.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | NM_015299.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400146889.hg.1 | TC1400006742.hg.1 | Clariom D TC |

KHNYN
Genomic Map