Assay Details
| Gene Symbol: AFDN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027906.1 | 1 - 2 | 1388 | 84 | ||
| GenBank mRNA | AB016898.1 | - | 1 - 2 | 1301 | 84 | |
| AK301419.1 | - | 1 - 2 | 1380 | 84 | ||
| AK307980.1 | - | 2 - 3 | 1042 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002319.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 494790 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 28848 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240578 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P334340 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 33535 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 32486 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3240578 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3240578 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P334340 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 9039 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P334340 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8130819 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130823 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130820 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130818 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130822 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8130821 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8130817 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 210409_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11736373_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11736373_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11736374_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11736374_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029105.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029105.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029105.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029101.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029101.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029101.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029109.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029109.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029109.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029108.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029108.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029108.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029110.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029110.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029103.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029103.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029103.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029104.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029104.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029104.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029102.hg.1 | TC06002319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029102.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06029102.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196081.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196080.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196080.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196080.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196080.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196080.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196085.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196090.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196090.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196090.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196090.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196090.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196094.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196094.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196094.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196086.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196086.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196086.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196086.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196086.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196088.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196091.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196078.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196078.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196078.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196078.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196078.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196082.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196082.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196082.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196082.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196082.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196087.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196087.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196087.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196087.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196087.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000414943.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000417244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013943.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196092.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196092.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196092.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196092.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196092.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | ENST00000359760.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | ENST00000425438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | NR_027906.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600196079.hg.1 | TC0600013943.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 210409_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 210409_at | - | - |
| Human Genome U133A 2.0 Array | 210409_PM_at | - | - |
| Human Genome U133A 2.0 Array | 210409_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw582114 | - | - |
| Human Genome U95 Set | 35608_at | - | - |
| Human Genome U95 Set | 35608_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 210409_at1 | - | - |
| Human Genome U133 Set | 210409_PM_at | - | - |
| Human Genome U133 Set | 210409_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030831 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030832 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030831 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030832 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 216087 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3242186 | AK091690.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242186 | CR600303.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242186 | NR_027906.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3242186 | AB016898.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1691374 | - | - |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242186 | AK091690.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242186 | CR600303.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242186 | NR_027906.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242186 | AB016898.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1691374 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1792570 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691374 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1691374 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1792570 | - | - |

AFDN-AS1
Genomic Map