Assay Details
| Gene Symbol: DIP2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014974.2 | NP_055789.1 | 34 - 35 | 4203 | 75 | |
| XM_005252426.3 | XP_005252483.1 | 35 - 36 | 4362 | 75 | ||
| XM_005252427.3 | XP_005252484.1 | 35 - 36 | 4273 | 75 | ||
| XM_005252428.4 | XP_005252485.1 | 36 - 37 | 4443 | 75 | ||
| XM_005252430.2 | XP_005252487.2 | 18 - 19 | 2453 | 75 | ||
| XM_011519428.2 | XP_011517730.1 | 36 - 37 | 4374 | 75 | ||
| XM_011519429.2 | XP_011517731.1 | 35 - 36 | 4342 | 75 | ||
| XM_011519430.2 | XP_011517732.1 | 35 - 36 | 4249 | 75 | ||
| XM_011519431.2 | XP_011517733.1 | 34 - 35 | 4219 | 75 | ||
| XM_011519432.1 | XP_011517734.1 | 35 - 36 | 4674 | 75 | ||
| XM_011519433.2 | XP_011517735.1 | 19 - 20 | 3482 | 75 | ||
| XM_017015977.1 | XP_016871466.1 | 19 - 20 | 2494 | 75 | ||
| GenBank mRNA | AB023151.2 | - | 34 - 35 | 4201 | 75 | |
| AK309715.1 | - | 18 - 19 | 2215 | 75 | ||
| BC063313.1 | - | 34 - 35 | 4203 | 30349267 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000961.hg.1
Set Membership: |
|

DIP2C
Genomic Map