Assay Details
| Gene Symbol: SMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323614.1 | NP_001310543.1 | 1 - 2 | 287 | 71 | |
| NM_001323615.1 | NP_001310544.1 | 1 - 2 | 287 | 71 | ||
| NM_015327.2 | NP_056142.2 | 1 - 2 | 226 | 71 | ||
| XM_017000843.1 | XP_016856332.1 | 1 - 2 | 233 | 71 | ||
| GenBank mRNA | AB029012.2 | - | 1 - 2 | 228 | 71 | |
| AB085691.1 | - | 1 - 2 | 154 | 71 | ||
| AK092183.1 | - | 1 - 2 | 202 | 71 | ||
| AY168922.1 | - | 1 - 2 | 80 | 71 | ||
| AY336728.1 | - | 1 - 2 | 77 | 71 | ||
| BC038296.1 | - | 1 - 2 | 227 | 4824487 | 71 | |
| CT000143.1 | - | 1 - 2 | 287 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003334.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017000843.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA export from nucleus
regulation of telomere maintenance
regulation of telomere maintenance via telomerase
regulation of dephosphorylation
protein binding
ubiquitin protein ligase binding
telomeric DNA binding
histone deacetylase binding
protein phosphatase 2A binding
DNA polymerase binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
mRNA export from nucleus
regulation of telomere maintenance
regulation of telomere maintenance via telomerase
regulation of dephosphorylation
protein binding
ubiquitin protein ligase binding
telomeric DNA binding
histone deacetylase binding
protein phosphatase 2A binding
DNA polymerase binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01051971.hg.1 | TC01003334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051971.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051971.hg.1 | ENST00000368267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051971.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051972.hg.1 | TC01003334.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051972.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051972.hg.1 | ENST00000368267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051972.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221122.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221122.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221122.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221122.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221122.hg.1 | TC0100015991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221123.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221123.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221123.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221123.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221123.hg.1 | TC0100015991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100221124.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221124.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221124.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100221124.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100221124.hg.1 | TC0100015991.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000361813.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000468993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000473643.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000476954.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | ENST00000489907.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015991.hg.1 | NM_015327.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 168706 | - | - |

SMG5
Genomic Map