Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020746.4 | NP_065797.2 | 2 - 3 | 291 | 69 | |
| NR_037921.1 | 2 - 3 | 291 | 69 | |||
| GenBank mRNA | AB033097.1 | - | 1 - 2 | 197 | 69 | |
| AB232371.1 | - | 1 - 2 | 191 | 69 | ||
| AK023799.1 | - | 2 - 3 | 247 | 69 | ||
| AK123956.1 | - | 2 - 3 | 252 | 69 | ||
| AK130992.1 | - | 2 - 3 | 212 | 69 | ||
| AK296897.1 | - | 2 - 3 | 256 | 69 | ||
| BC044952.1 | - | 2 - 3 | 225 | 5751684 | 69 | |
| BM834250.1 | - | 2 - 3 | 255 | 69 | ||
| DB219666.1 | - | 2 - 3 | 291 | 69 | ||
| DQ167126.1 | - | 1 - 2 | 119 | 69 | ||
| DQ174270.1 | - | 1 - 2 | 119 | 69 | ||
| DQ181928.1 | - | 1 - 2 | 119 | 69 | ||
| EF467324.1 | - | 1 - 2 | 119 | 69 | ||
| KC415005.1 | - | 1 - 2 | 119 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MAVS
Genomic Map