Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025584.hg.1 |
ENST00000270459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025584.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025584.hg.1 |
TC19001883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025581.hg.1 |
ENST00000270459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025581.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025581.hg.1 |
TC19001883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025583.hg.1 |
ENST00000270459.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025583.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025583.hg.1 |
TC19001883.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
TC1900011499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197462.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197464.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197464.hg.1 |
TC1900011499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197464.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
TC1900011499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197459.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197463.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197463.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197463.hg.1 |
TC1900011499.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197463.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197463.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000586787.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000587279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
ENST00000610935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011499.hg.1 |
NM_001002836.3 |
Refseq |