Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030713.hg.1 |
TC01001981.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030713.hg.1 |
ENST00000343783.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030713.hg.1 |
ENST00000355875.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030713.hg.1 |
ENST00000366541.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01030713.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100189840.hg.1 |
TC0100012200.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000366541.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000435549.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463012.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000463042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000476722.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000482234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000490065.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000491888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000493334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000496361.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
ENST00000497459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012200.hg.1 |
NM_006642.4 |
Refseq |