Assay Details
Gene Symbol: CASP16P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_132322.1 | 5 - 6 | 596 | 87 | ||
GenBank mRNA | AF098666.1 | - | 5 - 6 | 644 | 87 | |
BC111724.1 | - | 5 - 6 | 596 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403429 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892965 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892971 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16844982 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892972 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16844991 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892969 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30403428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892967 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09892950 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 237396 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 286333 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 319579 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 407766 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 42149 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 59478 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P740620 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20381998 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20381998 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12368290 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20381998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 234231_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147549.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147549.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147549.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147549.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147549.hg.1 | TC1600006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600147550.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147550.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147550.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147550.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147550.hg.1 | TC1600006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600147548.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147548.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147548.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147548.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147548.hg.1 | TC1600006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147545.hg.1 | TC1600006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600147546.hg.1 | TC1600006666.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000428155.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000571356.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000575497.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | ENST00000576807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006666.hg.1 | NR_132322.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 234231_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 234231_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 234231_at7 | - | - |
Human Genome U133 Set | 234231_at | - | - |
Human Genome U133 Set | 234231_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 192520 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789871 | AF098666.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1789871 | AF098666.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1789871 | AF098666.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1789871 | AF098666.1 | human_transcriptome |
