Assay Details
Gene Symbol: PAPPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020318.2 | NP_064714.2 | 13 - 14 | 5093 | 62 | |
XM_005245422.3 | XP_005245479.1 | 13 - 14 | 5140 | 62 | ||
XM_011509857.1 | XP_011508159.1 | 14 - 15 | 5010 | 62 | ||
XM_011509858.2 | XP_011508160.1 | 13 - 14 | 5353 | 62 | ||
XM_017002023.1 | XP_016857512.1 | 14 - 15 | 5191 | 62 | ||
XM_017002024.1 | XP_016857513.1 | 14 - 15 | 5238 | 62 | ||
XM_017002025.1 | XP_016857514.1 | 13 - 14 | 5344 | 62 | ||
GenBank mRNA | AB209369.1 | - | 5 - 6 | 631 | 62 | |
AF311940.1 | - | 12 - 13 | 3929 | 62 | ||
AF342989.1 | - | 13 - 14 | 5093 | 62 | ||
AJ278348.1 | - | 12 - 13 | 3892 | 62 | ||
BC152552.1 | - | 13 - 14 | 5081 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001534.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01023672.hg.1 | TC01001534.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01023672.hg.1 | ENST00000367662.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01023672.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367661.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000479836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000486075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | ENST00000493665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010713.hg.1 | NM_021936.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178708.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178708.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178708.hg.1 | ENST00000367662.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100178708.hg.1 | NM_020318.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100178708.hg.1 | TC0100010713.hg.1 | Clariom D TC |
