Assay Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020846.1 | NP_065897.1 | 3 - 4 | 844 | 76 | |
| XM_005248121.2 | XP_005248178.1 | 3 - 4 | 960 | 76 | ||
| GenBank mRNA | AB040891.1 | - | 3 - 4 | 814 | 76 | |
| AK026476.1 | - | 3 - 4 | 391 | 76 | ||
| BC031691.2 | - | 1 - 2 | 164 | 76 | ||
| BC040993.1 | - | 3 - 4 | 1123 | 5272534 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000278.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC031691.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04004007.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004007.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004007.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004007.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004006.hg.1 | TC04000278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004006.hg.1 | ENST00000264313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004006.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004006.hg.1 | ENST00000512093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04004006.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000505131.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000510595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007447.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000506375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150869.hg.1 | TC0400007447.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | ENST00000264313.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | ENST00000512093.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | NM_020846.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150870.hg.1 | TC0400007447.hg.1 | Clariom D TC |

SLAIN2
Genomic Map