Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000413030.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000426365.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000436950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000438115.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000449678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
ENST00000451350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007792.hg.1 |
NR_120420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006853.hg.1 |
ENST00000526867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006853.hg.1 |
ENST00000526867.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010107.hg.1 |
ENST00000528848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010107.hg.1 |
ENST00000528848.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010107.hg.1 |
NM_001256686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010107.hg.1 |
NM_001256686.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017909.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017909.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017909.hg.1 |
TC11001422.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100006853.hg.1 |
ENST00000526867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006853.hg.1 |
ENST00000526867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
ENST00000526064.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010105.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176739.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176739.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176739.hg.1 |
ENST00000227756.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176739.hg.1 |
NM_198516.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176739.hg.1 |
TC1100010105.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010107.hg.1 |
ENST00000528848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010107.hg.1 |
ENST00000528848.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010107.hg.1 |
NM_001256686.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010107.hg.1 |
NM_001256686.1 |
Refseq |