Assay Details
Gene Symbol: TFPI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271003.1 | NP_001257932.1 | 1 - 2 | 406 | 79 | |
GenBank mRNA | AK092499.1 | - | 1 - 2 | 168 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184386.hg.1 | TC0700011809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184391.hg.1 | TC0700011809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184390.hg.1 | TC0700011809.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000451238.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0700011809.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | ENST00000222543.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | ENST00000461482.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_001271003.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_001271004.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | NM_006528.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700184389.hg.1 | TC0700011809.hg.1 | Clariom D TC |
