Assay Details
| Gene Symbol: ADCK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174922.4 | NP_777582.4 | 3 - 4 | 345 | 77 | |
| XM_006716527.2 | XP_006716590.1 | 3 - 4 | 340 | 77 | ||
| XM_011516907.2 | XP_011515209.1 | 3 - 4 | 1058 | 77 | ||
| XM_011516909.1 | XP_011515211.1 | 3 - 4 | 345 | 77 | ||
| XM_017013174.1 | XP_016868663.1 | 3 - 4 | 383 | 77 | ||
| GenBank mRNA | AK092773.1 | - | 3 - 4 | 308 | 77 | |
| BC101659.1 | - | 3 - 4 | 285 | 8069165 | 77 | |
| BC143326.1 | - | 3 - 4 | 285 | 77 | ||
| BM146288.1 | - | 3 - 4 | 331 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000851.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_006716527.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011352.hg.1 | TC08000851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011352.hg.1 | ENST00000308860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011352.hg.1 | ENST00000526231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011352.hg.1 | ENST00000532190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011352.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163401.hg.1 | TC0800009254.hg.1 | Clariom D TC |

ADCK5
Genomic Map