Assay Details
| Gene Symbol: SDR16C5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318049.1 | NP_001304978.1 | 3 - 4 | 1188 | 108 | |
| NM_138969.3 | NP_620419.2 | 3 - 4 | 1188 | 108 | ||
| GenBank mRNA | AB083038.1 | - | 3 - 4 | 1094 | 108 | |
| AK057667.1 | - | 3 - 4 | 530 | 108 | ||
| AK095159.1 | - | 3 - 4 | 1188 | 108 | ||
| BC037219.1 | - | 3 - 4 | 535 | 5181472 | 108 | |
| BC064525.1 | - | 3 - 4 | 507 | 5752917 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001232.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | TC08001232.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | ENST00000303749.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | ENST00000396721.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | ENST00000538514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08015970.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010511.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171425.hg.1 | TC0800010511.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000303749.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000396721.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | ENST00000522671.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | NM_138969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800171424.hg.1 | TC0800010511.hg.1 | Clariom D TC |

SDR16C5
Genomic Map