Assay Details
Gene Symbol: DDX51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175066.3 | NP_778236.2 | 8 - 9 | 1287 | 102 | |
XM_011538256.1 | XP_011536558.1 | 8 - 9 | 1287 | 102 | ||
GenBank mRNA | AB593157.1 | - | 8 - 9 | 1307 | 102 | |
AK097078.1 | - | 8 - 9 | 1287 | 102 | ||
BC040185.2 | - | 8 - 9 | 1260 | 4801253 | 102 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12002148.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12028125.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12028125.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12028125.hg.1 | TC12002148.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200197639.hg.1 | TC1200012512.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
