Assay Details
Gene Symbol: AMER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152424.3 | NP_689637.3 | 1 - 2 | 174 | 86 | |
GenBank mRNA | AK097146.1 | - | 1 - 2 | 174 | 86 | |
DA939151.1 | - | 1 - 2 | 174 | 86 | ||
EF186024.1 | - | 1 - 2 | 35 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001095.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK097146.1,EF186024.1,NM_152424.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
positive regulation of protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
anatomical structure development
bone development
adipose tissue development
regulation of canonical Wnt signaling pathway
mesenchymal cell differentiation involved in kidney development
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
positive regulation of cellular protein catabolic process
beta-catenin destruction complex assembly
beta-catenin destruction complex disassembly
protein binding
phosphatidylinositol-4,5-bisphosphate binding
beta-catenin binding
beta-catenin destruction complex binding
Wnt signaling pathway
positive regulation of protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
anatomical structure development
bone development
adipose tissue development
regulation of canonical Wnt signaling pathway
mesenchymal cell differentiation involved in kidney development
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
positive regulation of cellular protein catabolic process
beta-catenin destruction complex assembly
beta-catenin destruction complex disassembly
protein binding
phosphatidylinositol-4,5-bisphosphate binding
beta-catenin binding
beta-catenin destruction complex binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X014889.hg.1 | TC0X001095.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014889.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014889.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014890.hg.1 | TC0X001095.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014890.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014890.hg.1 | ENST00000374869.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014890.hg.1 | ENST00000403336.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014890.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014892.hg.1 | TC0X001095.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014892.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014892.hg.1 | ENST00000374869.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014892.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014888.hg.1 | TC0X001095.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014888.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014888.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014891.hg.1 | TC0X001095.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014891.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014891.hg.1 | ENST00000374869.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X014891.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167562.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167564.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009877.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167559.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167559.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167559.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167559.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167559.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167563.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00167558.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167558.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167558.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167558.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167558.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167560.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | ENST00000330258.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | ENST00000374869.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | NM_152424.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00167561.hg.1 | TC0X00009877.hg.1 | Clariom D TC |
