Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182633.2 | NP_872439.2 | 4 - 5 | 1068 | 69 | |
| GenBank mRNA | AK097282.1 | - | 1 - 2 | 81 | 69 | |
| AL539604.3 | - | 4 - 5 | 831 | 69 | ||
| BC094796.1 | - | 4 - 5 | 890 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196785.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000484120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013361.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196783.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196781.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000482436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | ENST00000633730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196787.hg.1 | TC0700013361.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000411863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000429591.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | ENST00000466630.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | NM_182633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196780.hg.1 | TC0700013361.hg.1 | Clariom D TC |

ZNF713
Genomic Map