Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
TC01001493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000338920.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000367742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000392078.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000392080.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000412837.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
ENST00000451306.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023201.hg.1 |
NM_015172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000338920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000367742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000426496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
NM_015172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000338920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000367742.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
ENST00000426496.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
NM_015172.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178017.hg.1 |
TC0100010592.hg.1 |
Clariom D TC |