Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000436930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000457909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000622983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000623019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000623246.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000624922.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
NM_001293083.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000436930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000457909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000622983.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000623019.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000623246.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
ENST00000624922.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
NM_001293083.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200159138.hg.1 |
TC0200008512.hg.1 |
Clariom D TC |