Assay Details
Gene Symbol: SEC24A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021982.2 | NP_068817.1 | 21 - 22 | 3354 | 87 | |
XM_006714523.2 | XP_006714586.1 | 20 - 21 | 3555 | 87 | ||
XM_017008961.1 | XP_016864450.1 | 21 - 22 | 2999 | 87 | ||
XM_017008962.1 | XP_016864451.1 | 21 - 22 | 2771 | 87 | ||
GenBank mRNA | AJ131244.1 | - | 21 - 22 | 3017 | 87 | |
AK026545.1 | - | 8 - 9 | 979 | 87 | ||
AK304060.1 | - | 21 - 22 | 3343 | 87 | ||
AL137677.1 | - | 13 - 14 | 1633 | 87 | ||
BC009325.2 | - | 8 - 9 | 996 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000665.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
antigen processing and presentation of peptide antigen via MHC class I
intracellular protein transport
ER to Golgi vesicle-mediated transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of low-density lipoprotein particle receptor biosynthetic process
COPII vesicle coating
positive regulation of protein secretion
positive regulation of cholesterol homeostasis
molecular_function
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05009216.hg.1 | TC05000665.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05009216.hg.1 | ENST00000398844.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05009216.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000322887.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | ENST00000513123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_001252231.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008700.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159752.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159752.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159752.hg.1 | ENST00000398844.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500159752.hg.1 | NM_021982.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500159752.hg.1 | TC0500008700.hg.1 | Clariom D TC |
