| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004109.hg.1 |
ENST00000269214.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004109.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004109.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR18004109.hg.1 |
TC18000401.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
TC1800008231.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1800156209.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000269214.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000383276.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000580101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
ENST00000622333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800008231.hg.1 |
NM_052911.2 |
Refseq |