Assay Details
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110323.1 | 11 - 12 | 3166 | 95 | ||
GenBank mRNA | AL832066.1 | - | 11 - 12 | 3166 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927822 | LOC101927822 | -662.6kb | Anti-sense | Chr. 8 - 133886496 - 133902407 on NCBI Build GRCh38 | |
Downstream | ZFAT antisense RNA 1 | ZFAT-AS1 |
NCRNA00070, SAS-ZFAT, ZFAT-AS, ZFATAS |
32.7kb | Anti-sense | Chr. 8 - 134598071 - 134600689 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
