Assay Details
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL833686.1 | - | 3 - 4 | 1413 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000403496.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000423995.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000426597.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | ENST00000441044.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013603.hg.1 | NR_026850.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 100410 | - | - |
