Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016812.hg.1 |
TC09001340.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016812.hg.1 |
ENST00000356884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016812.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016812.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016812.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173580.hg.1 |
TC0900010808.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000356884.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
ENST00000375512.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_001003800.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010808.hg.1 |
NM_015250.3 |
Refseq |