Assay Details
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246818.1 | XP_005246875.1 | 3 - 4 | 447 | 105 | |
XM_005246819.1 | XP_005246876.1 | 2 - 3 | 195 | 105 | ||
XM_006712720.3 | XP_006712783.1 | 3 - 4 | 234 | 105 | ||
XM_011511707.2 | XP_011510009.1 | 4 - 5 | 575 | 105 | ||
XM_011511708.1 | XP_011510010.1 | 3 - 4 | 509 | 105 | ||
XM_011511709.2 | XP_011510011.1 | 3 - 4 | 277 | 105 | ||
GenBank mRNA | AB209866.1 | - | 2 - 3 | 6369 | 105 | |
AF021834.1 | - | 3 - 4 | 391 | 105 | ||
AK313562.1 | - | 2 - 3 | 359 | 105 | ||
AU136083.1 | - | 3 - 4 | 538 | 105 | ||
BC015514.1 | - | 2 - 3 | 361 | 3902987 | 105 | |
DB078397.1 | - | 2 - 3 | 417 | 105 | ||
J03225.1 | - | 2 - 3 | 253 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800006681.hg.1 | ENST00000579385.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 126759 | - | - |
