Assay Details
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246818.1 | XP_005246875.1 | 7 - 8 | 949 | 68 | |
XM_005246819.1 | XP_005246876.1 | 6 - 7 | 697 | 68 | ||
XM_006712720.3 | XP_006712783.1 | 7 - 8 | 736 | 68 | ||
XM_011511707.2 | XP_011510009.1 | 8 - 9 | 1077 | 68 | ||
XM_011511708.1 | XP_011510010.1 | 7 - 8 | 1011 | 68 | ||
XM_011511709.2 | XP_011510011.1 | 7 - 8 | 779 | 68 | ||
GenBank mRNA | AB209866.1 | - | 5 - 6 | 7465 | 68 | |
AK313562.1 | - | 6 - 7 | 861 | 68 | ||
J03225.1 | - | 6 - 7 | 755 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000233156.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000339091.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000392365.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000409676.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000417013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000420747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000421427.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000426055.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000435414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000437725.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000453013.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481132.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | ENST00000481389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_001032281.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015194.hg.1 | NM_006287.5 | Refseq |
