| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008648.hg.1 |
TC04000590.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008648.hg.1 |
ENST00000310836.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008648.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008648.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008648.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008645.hg.1 |
TC04000590.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008645.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04008645.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158250.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158250.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158250.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158250.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158250.hg.1 |
TC0400008483.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400158254.hg.1 |
TC0400008483.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000310836.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000394511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
ENST00000507710.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_001128174.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008483.hg.1 |
NM_003360.4 |
Refseq |