Assay Details
| Gene Symbol: INPP5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321042.1 | NP_001307971.1 | 10 - 11 | 1456 | 74 | |
| NM_005539.4 | NP_005530.3 | 13 - 14 | 1403 | 74 | ||
| XM_017016204.1 | XP_016871693.1 | 12 - 13 | 1057 | 74 | ||
| GenBank mRNA | BC052265.1 | - | 10 - 11 | 830 | 74 | |
| BC062300.1 | - | 10 - 11 | 830 | 74 | ||
| BC096280.2 | - | 14 - 15 | 1147 | 40006844 | 74 | |
| BC096281.2 | - | 13 - 14 | 1066 | 40006846 | 74 | |
| X77567.1 | - | 13 - 14 | 1184 | 74 | ||
| Z31695.1 | - | 13 - 14 | 1039 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000946.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | ENST00000368594.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | ENST00000416326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | ENST00000432898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | ENST00000536599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011676.hg.1 | TC10000946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | ENST00000368594.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | ENST00000416326.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | ENST00000432898.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | ENST00000536599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10011675.hg.1 | TC10000946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000342652.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368593.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000423490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000487614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | ENST00000498337.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009414.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164408.hg.1 | TC1000009414.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | ENST00000368594.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | ENST00000445580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | NM_005539.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000164409.hg.1 | TC1000009414.hg.1 | Clariom D TC |

INPP5A
Genomic Map