Assay Details
| Gene Symbol: LOC105373876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135525.1 | 1 - 2 | 230 | 100 | ||
| GenBank mRNA | BC031974.1 | - | 1 - 2 | 235 | 100 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105373876
Genomic Map