Assay Details
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U92995.1 | - | 1 - 2 | 49 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000337672.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000431606.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000528205.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | ENST00000534677.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010905.hg.1 | NM_153450.2 | Refseq |
