Assay Details
| Gene Symbol: DGCR5 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110533.1 | 5 - 6 | 862 | 108 | |||
| GenBank mRNA | X91348.1 | - | LIT1227 DGCR5 |
5 - 6 | 834 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000399539.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000399539.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000399539.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000421572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000421572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000421572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000421572.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000424407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000438934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000440005.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000537283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000540720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000540720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | ENST00000609630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_002733.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_024159.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_045121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006577.hg.1 | NR_110533.1 | Refseq |

DGCR5
Genomic Map