Assay Details
| Gene Symbol: TYSND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040273.2 | NP_001035363.1 | 1 - 2 | 1320 | 83 | |
| GenBank mRNA | BC030242.1 | - | 1 - 2 | 1236 | 5176098 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | ENST00000287078.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | ENST00000335494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | ENST00000494143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | NM_001040273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | NM_173555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017183.hg.1 | TC10001360.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000337198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000347770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518353.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000518940.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000520402.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000521536.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000522311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | ENST00000523071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_001301668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_015878.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012447.hg.1 | NM_148174.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | ENST00000287078.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | ENST00000335494.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | ENST00000494143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | NM_001040273.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | NM_173555.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10017185.hg.1 | TC10001360.hg.1 | HTA TC |

TYSND1
Genomic Map