Assay Details
| Gene Symbol: TDRD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527471.2 | XP_011525773.1 | 16 - 17 | 1811 | 100 | |
| XM_011527472.2 | XP_011525774.1 | 16 - 17 | 1811 | 100 | ||
| XM_011527473.2 | XP_011525775.1 | 16 - 17 | 1811 | 100 | ||
| XM_011527474.2 | XP_011525776.1 | 15 - 16 | 1691 | 100 | ||
| XM_011527475.2 | XP_011525777.1 | 16 - 17 | 1811 | 100 | ||
| XM_011527476.2 | XP_011525778.1 | 16 - 17 | 1811 | 100 | ||
| XM_017027458.1 | XP_016882947.1 | 16 - 17 | 1811 | 100 | ||
| XM_017027459.1 | XP_016882948.1 | 6 - 7 | 794 | 100 | ||
| GenBank mRNA | BC049000.1 | - | 16 - 17 | 2051 | 100 | |
More Information
Set Membership: |
|

TDRD12
Genomic Map