Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019403.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019403.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019403.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019403.hg.1 |
TC19001444.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
ENST00000484218.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_001244847.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010478.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
NM_207392.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
TC1900010478.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
ENST00000338897.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
ENST00000479340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900185595.hg.1 |
NM_207392.2 |
Refseq |