Assay Details
| Gene Symbol: STARD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017025553.1 | XP_016881042.1 | 1 - 2 | 237 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017025553.1 transcripts.
Set Membership: |
|

STARD6
Genomic Map