Assay Details
Gene Symbol: LINC01346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040065.1 | 1 - 2 | 83 | 87 | ||
GenBank mRNA | AK124708.1 | - | 1 - 2 | 78 | 87 | |
DB288420.1 | - | 1 - 2 | 130 | 87 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000076.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012595 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021393 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012570 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012585 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012614 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021365 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012563 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021387 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012588 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021390 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012594 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021404 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012581 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021397 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012598 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021360 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012610 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016008 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021388 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016010 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012584 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021380 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021403 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15016009 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012579 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20021353 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012604 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012578 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00012609 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 81376 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 502980 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 266210 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 524733 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 425572 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P932339 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P932339 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 10424 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13103 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P932339 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P932339 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P932339 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00004032 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00004032 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 69238 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 56736 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 75282 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00004045 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00004032 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00004040 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00004032 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00004045 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1566735_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1566737_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01001218.hg.1 | TC01000076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001218.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001218.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001218.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01001219.hg.1 | TC01000076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01001219.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001219.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01001219.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100006631.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100145918.hg.1 | TC0100006631.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000412674.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | ENST00000456897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | NR_040065.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100145917.hg.1 | TC0100006631.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 1566735_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566735_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566737_at | - | - |
Human Genome U133 Plus 2.0 Array | 1566737_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566735_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1566737_at11 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3228056 | DB288420.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3228056 | NR_040065.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3228056 | AK124708.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3233656 | DB288420.1 | human_transcriptome |
